Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 4/CHRNA4 Rabbit anti-Human, Mouse, Rat, Clone: 1C2H2, Novus Biologicals™
SDP

Catalog No. NB165617 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB165617 20 μg
NB165616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB165617 Supplier Novus Biologicals Supplier No. NBP31526020UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nicotinic Acetylcholine R alpha 4/CHRNA4 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 4/CHRNA4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Nicotinic Acetylcholine R alpha 4/CHRNA4
Applications Western Blot
Classification Monoclonal
Clone 1C2H2
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BFNC, cholinergic receptor, nicotinic, alpha 4, cholinergic receptor, nicotinic, alpha polypeptide 4, EBN, EBN1, ENFL1, FLJ95812, NACHR, NACHRA4, NACRA4, neuronal acetylcholine receptor subunit alpha-4, neuronal nicotinic acetylcholine receptor alpha-4 subunit
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 500-600 of human Nicotinic Acetylcholine R alpha 4/CHRNA4 (P43681).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 1137
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.