Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine R alpha 7/CHRNA7 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB327239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327239 Supplier Novus Biologicals Supplier No. NBP299069MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Nicotinic Acetylcholine R alpha 7/CHRNA7 Polyclonal antibody specifically detects Nicotinic Acetylcholine R alpha 7/CHRNA7 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Nicotinic Acetylcholine R alpha 7/CHRNA7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias a7 nicotinic acetylcholine receptor, alpha 7 neuronal nicotinic acetylcholine receptor, alpha-7 nicotinic cholinergic receptor subunit, cholinergic receptor, nicotinic, alpha 7, cholinergic receptor, nicotinic, alpha polypeptide 7, CHRNA7-2, NACHRA7, neuronal acetylcholine receptor protein, alpha-7 chain, neuronal acetylcholine receptor subunit alpha-7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nicotinic Acetylcholine R alpha 7/CHRNA7.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1139
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.