Learn More
Nicotinic Acetylcholine Receptor beta 4 Antibody (1H11F2) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Nicotinic Acetylcholine Receptor beta 4 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1H11F2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | cholinergic receptor, nicotinic, beta 4, cholinergic receptor, nicotinic, beta polypeptide 4, neuronal acetylcholine receptor subunit beta-4, neuronal nicotinic receptor beta 4 subunit |
| Gene Symbols | CHRNB4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.