Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nicotinic Acetylcholine Receptor epsilon Antibody, Novus Biologicals™
SDP

Catalog No. NBP226108 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226108 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP226108 Supplier Novus Biologicals Supplier No. NBP226108
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Nicotinic Acetylcholine Receptor epsilon Polyclonal specifically detects Nicotinic Acetylcholine Receptor epsilon in Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen Nicotinic Acetylcholine Receptor epsilon
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Alias acetylcholine receptor subunit epsilon, AchR epsilon subunit, ACHREFCCMS, cholinergic receptor, nicotinic, epsilon, cholinergic receptor, nicotinic, epsilon polypeptide, CMS1D, CMS1E, CMS2A, SCCMS
Gene Symbols CHRNE
Host Species Goat
Immunogen Peptide with sequence C-DQEATGEEVSD corresponding to internal region according to NP_000071.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1145
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.