Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nidogen-1/Entactin Antibody (ELM1), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB382535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382535 Supplier Novus Biologicals Supplier No. NBP308237JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Nidogen-1/Entactin Monoclonal antibody specifically detects Nidogen-1/Entactin in Mouse, Guinea Pig samples. It is validated for ELISA

Specifications

Antigen Nidogen-1/Entactin
Applications ELISA
Classification Monoclonal
Clone ELM1
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias enactin, Entactin, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1
Host Species Rat
Immunogen Murine EHS laminin preparation
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4811
Target Species Mouse, Guinea Pig
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.