Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann-Pick C1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB400148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400148 0.1 mL
NB400148SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400148 Supplier Novus Biologicals Supplier No. NB400148
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 42 publications

Niemann-Pick C1 Polyclonal specifically detects Niemann-Pick C1 in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy, Knockout Validated, Knockdown Validated.

Specifications

Antigen Niemann-Pick C1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:3000. Use reported in scientific literature (PMID 24209575), Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence 1:250. Use reported in scientific literature (PMID 24209575), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 18216017), Immunohistochemistry-Paraffin 5-10 ug/ml, Electron Microscopy reported in scientific literature (PMID 21051527), Knockout Validated, KnockDown Validated
Formulation PBS with 0.1% Sodium Azide
Gene Accession No. O15118
Gene Alias FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1
Gene Symbols NPC1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4864
Test Specificity Human, mouse, rat and Chinese hamster and primate (PMID 22212234). Porcine reactivity reported in scientific literature (PMID: 21051527).
Target Species Human, Mouse, Rat, Pig, Chinese Hamster, Hamster, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.