Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann-Pick C1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB359400 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359400 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359400 Supplier Novus Biologicals Supplier No. NB400148PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Niemann-Pick C1 Polyclonal antibody specifically detects Niemann-Pick C1 in Human, Mouse, Rat, Porcine, Chinese Hamster, Hamster, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunomicroscopy, KnockDown, KnockDown

Specifications

Antigen Niemann-Pick C1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Electron Microscopy, Gene Knock-Out, KnockDown
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy, Knockout Validated, Knockdown Validated
Formulation PBS
Gene Alias FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1
Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4864
Target Species Human, Mouse, Rat, Pig, Chinese Hamster
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.