Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann-Pick type C1 Like-1 Antibody, Novus Biologicals™
SDP

Catalog No. NB400127SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400127SS 0.025 mL
NB400127 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400127SS Supplier Novus Biologicals Supplier No. NB400127SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 15 publications

Niemann-Pick type C1 Like-1 Polyclonal specifically detects Niemann-Pick type C1 Like-1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.

Specifications

Antigen Niemann-Pick type C1 Like-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, ELISA reported in scientific literature (PMID 21051527), Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Electron Microscopy reported in scientific literature (PMID 21051527), KnockDown Validated reported in scientific literature (PMID 35023619)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
Gene Alias Niemann-Pick C1-like protein 1, NPC1 (Niemann-Pick disease, type C1, gene)-like 1, NPC11L1
Gene Symbols NPC1L1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 1000-1100). [UniProt# Q6T3U3]
Molecular Weight of Antigen 140 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 29881
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.