Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Niemann-Pick type C1 Like-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 15 publications
$206.50 - $499.50
Specifications
| Antigen | Niemann-Pick type C1 Like-1 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, ELISA reported in scientific literature (PMID 21051527), Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Electron Microscopy reported in scientific literature (PMID 21051527), KnockDown Validated reported in scientific literature (PMID 35023619) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400127SS
![]() |
Novus Biologicals
NB400127SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400127
![]() |
Novus Biologicals
NB400127 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Niemann-Pick type C1 Like-1 Polyclonal specifically detects Niemann-Pick type C1 Like-1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| Niemann-Pick type C1 Like-1 | |
| Western Blot 2 ug/ml, ELISA reported in scientific literature (PMID 21051527), Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Electron Microscopy reported in scientific literature (PMID 21051527), KnockDown Validated reported in scientific literature (PMID 35023619) | |
| Polyclonal | |
| Rabbit | |
| Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| 29881 | |
| A synthetic peptide made to an internal region of rat Niemann-Pick type C1 Like-1 (between residues 1000-1100). [UniProt# Q6T3U3] | |
| Primary | |
| Store at 4C. Do not freeze. |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig | |
| Niemann-Pick C1-like protein 1, NPC1 (Niemann-Pick disease, type C1, gene)-like 1, NPC11L1 | |
| NPC1L1 | |
| IgG | |
| Affinity Purified | |
| 140 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title