Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann-Pick type C1 Like-1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB096429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096429 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB096429 Supplier Novus Biologicals Supplier No. NBP289182AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Niemann-Pick type C1 Like-1 Polyclonal specifically detects Niemann-Pick type C1 Like-1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Niemann-Pick type C1 Like-1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias Niemann-Pick C1-like protein 1, NPC1 (Niemann-Pick disease, type C1, gene)-like 1, NPC11L1
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the rat NPC1L1 protein (between amino acids 185-240) [UniProt Q6T3U3]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29881
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.