Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niemann-Pick type C2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB323172 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323172 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323172 Supplier Novus Biologicals Supplier No. NBP299920APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Niemann-Pick type C2 Polyclonal antibody specifically detects Niemann-Pick type C2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Niemann-Pick type C2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EDDM1, epididymal secretory protein E1, He1, HE1epididymal protein 1, Human epididymis-specific protein 1, MGC1333, Niemann-Pick disease type C2 protein, Niemann-Pick disease, type C2, NP-C2, tissue-specific secretory protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Niemann-Pick type C2 (Accession#: NP_006423.1; Met1-Leu151)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10577
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.