Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1002578 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002578 Supplier Novus Biologicals Supplier No. NB1002578
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIF1 Polyclonal specifically detects NIF1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen NIF1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_071378.1
Gene Alias DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein
Gene Symbols ZNF335
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human NRC-Interacting Factor 1 using the numbering given in entry NP_071378.1 (GeneID 63925).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63925
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Dog, Horse, Rabbit, Guinea pig, Pig, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.