Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB292301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292301 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292301 Supplier Novus Biologicals Supplier No. NB1002578PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIF1 Polyclonal specifically detects NIF1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen NIF1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry, Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias DKFZp586G1122, Hzf, HZFzinc finger protein 385, RZFHematopoietic zinc finger protein, ZFP385, zinc finger protein 385A, ZNF385Retinal zinc finger protein
Host Species Rabbit
Immunogen Maps to a region between residue 300 and 350 of human NRC-Interacting Factor 1 using the numbering given in entry NP_071378.1 (GeneID 63925).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63925
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.