Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIF3L1 Antibody (OTI5A7), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP272988FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272988FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272988FR Supplier Novus Biologicals Supplier No. NBP272988FR

Mouse Monoclonal Antibody

NIF3L1 Monoclonal specifically detects NIF3L1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NIF3L1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI5A7
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALS2CR1, candidate 1, MDS015, NIF3 NGG1 interacting factor 3-like 1 (S. pombe), NIF3-like protein 1, S.pombe homolog)-like 1
Gene Symbols NIF3L1
Host Species Mouse
Immunogen Full length recombinant protein of human NIF3L1 (NP_068596) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 60491.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.