Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIFK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | NIFK |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056661T
![]() |
Novus Biologicals
NB10056661SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056661
![]() |
Novus Biologicals
NB10056661 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
NIFK Polyclonal specifically detects NIFK in Human, Mouse samples. It is validated for Western Blot.Specifications
NIFK | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
Q9BYG3 | |
84365 | |
This antibody was developed against a synthetic peptide corresponding to amino acids 75-90 of mouse NIFK. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67 | |
MKI67IP | |
IgG | |
Protein G purified | |
In NIH 3T3, a 34 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title