Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIFK Antibody (18E148), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB339511 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB339511 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB339511 Supplier Novus Biologicals Supplier No. NB10056590MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NIFK Monoclonal antibody specifically detects NIFK in Human samples. It is validated for Western Blot

Specifications

Antigen NIFK
Applications Western Blot
Classification Monoclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias hNIFK, MKI67 (FHA domain) interacting nucleolar phosphoprotein, MKI67 FHA domain-interacting nucleolar phosphoprotein, NIFKNOPP34, Nopp34, Nucleolar phosphoprotein Nopp34, Nucleolar protein interacting with the FHA domain of pKI-67
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to a portion of human NIFK between amino acids 100-200.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 84365
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.