Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nigericin sodium salt
GREENER_CHOICESDP

Catalog No. 431210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-1210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 43-1210 Supplier Tocris Bioscience™ Supplier No. 4312/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective K+ ionophore

Potassium ionophore, exchanges K+ for H+ across biological membranes, in a similar manner to Valinomycin (Cat. No. 3373). Stimulates mitochondral ATPase activity and disrupts membrane potential. Also acts as an ionophore for Pb2+ with no activity with other divalent cations. Antibiotic derived from Streptomyces hygroscopius.

Chemical Identifiers

CAS 28643-80-3
Molecular Formula C40H67NaO11

Specifications

Quantity 10 mg
Formula Weight 746.94
Purity >98%
Molecular Weight (g/mol) 746.955
Chemical Name or Material Nigericin sodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.