Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIK/MAP3K14 Rabbit anti-Human, Mouse, Rat, Canine, Equine, Hamster, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1507480.025MG
Description
NIK/MAP3K14 Polyclonal specifically detects NIK/MAP3K14 in Human, Mouse, Rat, Canine, Equine, Hamster samples. It is validated for Western Blot, Simple Western.Specifications
NIK/MAP3K14 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 50% glycerol, 1% BSA. with 0.02% Thimerosal | |
EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK | |
Rabbit | |
104 kDa | |
0.025 mg | |
Apoptosis, Cancer, Protein Kinase, Signal Transduction | |
9020 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/mL | |
Western Blot 0.5 - 4 ug/mL, Simple Western | |
Q99558 | |
MAP3K14 | |
Synthetic peptide surrounding amino acid 927of human NIK | |
Affinity Purified | |
RUO | |
Primary | |
Hamster, Canine, Mouse, Human, Rat, Equine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction