Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIK/MAP3K14 Rabbit anti-Human, Mouse, Rat, Canine, Equine, Hamster, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | NIK/MAP3K14 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL, Simple Western |
Applications | Western Blot |
Classification | Polyclonal |
Description
NIK/MAP3K14 Polyclonal specifically detects NIK/MAP3K14 in Human, Mouse, Rat, Canine, Equine, Hamster samples. It is validated for Western Blot, Simple Western.Specifications
NIK/MAP3K14 | |
Western Blot 0.5 - 4 ug/mL, Simple Western | |
Polyclonal | |
Rabbit | |
Apoptosis, Cancer, Protein Kinase, Signal Transduction | |
PBS (pH 7.2), 50% glycerol, 1% BSA. with 0.02% Thimerosal | |
EC 2.7.11.25, FTDCR1B, HS, HsNIK, HSNIKserine/threonine protein-kinase, mitogen-activated protein kinase kinase kinase 14, NIKNF-kappa-beta-inducing kinase, Serine/threonine-protein kinase NIK | |
MAP3K14 | |
IgG | |
Affinity Purified | |
104 kDa |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Hamster, Canine, Mouse, Human, Rat, Equine | |
Q99558 | |
9020 | |
Synthetic peptide surrounding amino acid 927of human NIK | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title