Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninein Antibody, Novus Biologicals™
SDP

Catalog No. NB10074631 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074631 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074631 Supplier Novus Biologicals Supplier No. NB10074631
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ninein Polyclonal specifically detects Ninein in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen Ninein
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q8N4C6
Gene Alias Glycogen synthase kinase 3 beta-interacting protein, GSK3B-interacting protein, hNinein, KIAA1565, ninein, ninein (GSK3B interacting protein), ninein centrosomal protein
Gene Symbols NIN
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1850 and 1900 of human Ninein using the numbering given in entry NP_891991.1 (GeneID 51199).
Molecular Weight of Antigen 243 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51199
Test Specificity Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Gorilla, Chimpanzee and Northern white-cheeked gibbon.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.