Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninhydrin, 99%

Catalog No. AAA1040930
Click to view available options
Quantity:
10 g
25 g
50 g
250 g

Catalog No. AAA1040930

Supplier: Thermo Scientific Chemicals A1040930

Only null left
Add to Cart

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol

Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ninhydrin is used for the determination of amino groups in amino acids, peptides and proteins. It finds application in forensic science for the analysis of latent fingerprints on porous surfaces such as paper. It is also used in thin layer chromatography (TLC) to monitor the progress of chemical reactions. In Kaiser test, it plays an important role to monitor the deprotection in solid phase peptide synthesis.

Solubility
Soluble in water, acetone, ethanol, and methanol. Slightly soluble in chloroform and ether.

Notes
Light Sensitive. Incompatible with strong oxidizing agents, amines, alkalis and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
MDL Number MFCD00003791
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
Synonym ninhydrin, ninhydrin hydrate, 1,2,3-indantrione monohydrate, ninhydrine, 2,2-dihydroxy-1h-indene-1,3 2h-dione, 2,2-dihydroxy-1,3-indandione, triketohydrindene hydrate, 1h-indene-1,3 2h-dione, 2,2-dihydroxy, 1,2,3-indantrione, 2-hydrate, 2,2-dihydroxyindane-1,3-dione
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Melting Point 251°C (decomposition)
Assay Percent Range 99%
Quantity 250 g
Beilstein 1910963
Sensitivity Light sensitive
Merck Index 14,6554
Solubility Information Soluble in water,acetone,ethanol,and methanol. Slightly soluble in chloroform and ether.
Formula Weight 178.14
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ninhydrin
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-618-1
RTECSNumber NK5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.