Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ninhydrin, Calbiochem™,

Catalog No. nx04032 Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 g
1 kg
Packaging:
Plastic Bottle

Suitable for biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-47-2
Molecular Formula C9H6O4
Molecular Weight (g/mol) 178.143
InChI Key FEMOMIGRRWSMCU-UHFFFAOYSA-N
PubChem CID 10236
ChEBI CHEBI:86374
IUPAC Name 2,2-dihydroxyindene-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(C2=O)(O)O

Specifications

Percent Purity >99%
Packaging Plastic Bottle
pH 4.6 to 5.6
Quantity 10 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.