Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NINJ2 Mouse anti-Human, Clone: 5C5B2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | NINJ2 |
---|---|
Clone | 5C5B2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6635
![]() ![]() |
Proteintech Group Inc
662021IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Homophilic cell adhesion molecule that promotes axonal growth. May play a role in nerve regeneration and in the formation and function of other tissues.Specifications
NINJ2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Nerve injury induced protein 2, NINJ2, ninjurin 2 | |
NINJ2 | |
IgG2b | |
Protein A | |
Antibody |
5C5B2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NZG7 | |
4815 | |
NINJ2 Fusion Protein Ag5343 | |
Primary | |
-20°C | |
NINJ2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title