Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ninjurin-1 Antibody (758926), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312148 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312148 Supplier Novus Biologicals Supplier No. FAB51051APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ninjurin-1 Monoclonal specifically detects Ninjurin-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Ninjurin-1
Applications Flow Cytometry
Classification Monoclonal
Clone 758926
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJURIN, ninjurin 1, ninjurin-1
Host Species Mouse
Immunogen E. coli-derived recombinant human Ninjurin-1, Asp2-Val81, Accession # Q92982
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4814
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.