Learn More
Niobium(IV) oxide, 99+% (metals basis)
Description
Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 10 g |
| Assay Percent Range | >99% (metals basis) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 124.91 |
| Density | 5.9 g/mL |
| Chemical Name or Material | Niobium(IV) oxide |
Safety and Handling
| EINECSNumber | 234-809-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.