Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen NIP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17988320
SDP
View Documents
Novus Biologicals
NBP17988320UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP179883
SDP
View Documents
Novus Biologicals
NBP179883
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

NIP Polyclonal specifically detects NIP in Human samples. It is validated for Western Blot.
Specifications

Specifications

NIP
Polyclonal
Rabbit
NP_653166
90527
The specific Immunogen is proprietary information. Peptide sequence PEEGGLLSPRYRSMADSPKSQDIPLSEASSTKAYYRPRRLSLVPADVRGL.
Primary
Western Blot
Unconjugated
RUO
Dual oxidase activator 1, dual oxidase maturation factor 1, dual oxidase maturation factor 1 delta, dual oxidase maturation factor 1 gamma, FLJ32334, homolog of Drosophila Numb-interacting protein, mol, NIPdual oxidase maturation factor 1 alpha, Numb-interacting protein, NUMBIPdual oxidase maturation factor 1 beta
DUOXA1
IgG
53 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.