Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPSNAP1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB357138 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357138 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357138 Supplier Novus Biologicals Supplier No. NBP176888MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIPSNAP1 Polyclonal antibody specifically detects NIPSNAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NIPSNAP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias nipsnap homolog 1 (C. elegans), NIPSNAP, C. elegans, homolog 1,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 1, NipSnap1, protein NipSnap homolog 1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide from near the center of human NIPSNAP. The immunogen is located within amino acids 130 - 180 of NIPSNAP. Amino Acid Squence: CMNKLKNNKEYLEFRRERT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8508
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.