Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIPSNAP3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | NIPSNAP3A |
---|---|
Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NIPSNAP3A Polyclonal antibody specifically detects NIPSNAP3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NIPSNAP3A | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Neuroscience | |
PBS (pH 7.2), 40% Glycerol | |
DKFZp564D177, FLJ13953, HSPC299, MGC14553, nipsnap homolog 3A (C. elegans), NipSnap3A, NipSnap4, protein NipSnap homolog 3A, Protein NipSnap homolog 4, Target for Salmonella secreted protein C, TassC | |
This antibody was developed against Recombinant Protein corresponding to amino acids: VHVLWWNESADSRAAGRHKSHEDPRVVAAVRESVNYLVSQQNMLLIP | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Q9BS92 | |
25934 | |
IgG | |
Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title