Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIPSNAP3B Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB355951 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355951 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355951 Supplier Novus Biologicals Supplier No. NBP177341PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIPSNAP3B Polyclonal antibody specifically detects NIPSNAP3B in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NIPSNAP3B
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias FLJ11275, nipsnap homolog 3B (C. elegans), NIPSNAP3, NipSnap3B, protein NipSnap homolog 3B, SNAP1
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human NIPSNAP3B. The immunogen is located within amino acids 40 - 90 of NIPSNAP3B. Amino Acid Squence: AFMENLKKNIHLRTSY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 55335
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.