Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIRF Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB318043 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318043 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318043 Supplier Novus Biologicals Supplier No. NBP298956APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NIRF Polyclonal antibody specifically detects NIRF in Human samples. It is validated for Western Blot

Specifications

Antigen NIRF
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NIRF.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 115426
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.