Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NIRF Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$244.00
Specifications
| Antigen | NIRF |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019689
![]() |
Novus Biologicals
NBP2414160.025MG |
0.025 mg |
Each for $244.00
|
|
|||||
NBP24141650
![]() |
Novus Biologicals
NBP24141650UG |
50 μg | N/A | N/A | N/A | ||||
Description
NIRF Polyclonal specifically detects NIRF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| NIRF | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Rabbit | |
| PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal | |
| DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2 | |
| UHRF2 | |
| IgG | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human | |
| Q96PU4 | |
| 115426 | |
| Recombinant protein of human UHRF2. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title