Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NIRF Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB019689 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019689 0.025 mg
NBP24141650 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019689 Supplier Novus Biologicals Supplier No. NBP2414160.025MG

Rabbit Polyclonal Antibody

NIRF Polyclonal specifically detects NIRF in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NIRF
Applications Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. Q96PU4
Gene Alias DKFZp434B0920, DKFZp686G0837, E3 ubiquitin-protein ligase UHRF2, EC 6.3.2, EC 6.3.2.-, MGC33463, NIRFUbiquitin-like-containing PHD and RING finger domains protein 2, Np95/ICBP90-like RING finger protein, Np95-like RING finger protein, Nuclear protein 97, Nuclear zinc finger protein Np97, RNF107RING finger protein 107, Ubiquitin-like PHD and RING finger domain-containing protein 2, ubiquitin-like with PHD and ring finger domains 2, ubiquitin-like, containing PHD and RING finger domains, 2, URF2
Gene Symbols UHRF2
Host Species Rabbit
Immunogen Recombinant protein of human UHRF2.
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 115426
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.