Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595756
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA595756 100 μL
1 options

Catalog No. PIPA595756

Supplier: Invitrogen™ PA595756

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLGFITRPPH RFLSLLCPGL RIPQLSVLCA QPRPRAMAIS SSSCELPLVA VCQVTSTPDK QQNFKTCAEL VREAARLGAC LAFLPEAFDF IARDPAETLH LSEPLGGKLL EEYTQLAREC GLWLSLGGFH ERGQDWEQTQ KIYNCHVLLN; Positive Samples: Rat liver; Cellular Location: Cytoplasm, Mitochondrion.

Plays a role in cell growth and apoptosis: loss of expression promotes cell growth and resistance to DNA damage stress. Has tumor suppressor properties that enhances the apoptotic responsiveness in cancer cells; this effect is additive to the tumor suppressor activity of FHIT. It is also a negative regulator of primary T-cells. Has apparently no omega-amidase activity such as NIT2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NIT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NIT1
Gene Accession No. Q7TQ94
Gene Alias AI255805; Deaminated glutathione amidase; dGSH amidase; ESTM30; MGC57670; Nit1; nitrilase 1; nitrilase homolog 1; rCG_20134; W57327
Gene Symbols NIT1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human NIT1 (NP_0011720211).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289222
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.