Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilase 1 Antibody (OTI3A11), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N272999APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272999APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272999APC7 Supplier Novus Biologicals Supplier No. NBP272999APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nitrilase 1 Monoclonal specifically detects Nitrilase 1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Nitrilase 1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3A11
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.5, MGC57670, nitrilase 1, nitrilase homolog 1
Gene Symbols NIT1
Host Species Mouse
Immunogen Full length recombinant protein of human Nitrilase 1 (NP_005591) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4817.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.