Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotryptophan Antibody (2D12), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB259354647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259354647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259354647 Supplier Novus Biologicals Supplier No. NBP259354AF647

Mouse Monoclonal Antibody

Nitrotryptophan Monoclonal specifically detects Nitrotryptophan in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Nitrotryptophan
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2D12
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Test Specificity Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins.
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.