Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Antibody (39B6), PE, Novus Biologicals™
SDP

Catalog No. NBT96877PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBT96877PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBT96877PE Supplier Novus Biologicals Supplier No. NB11096877PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Rat, Non-species specific samples. It is validated for Flow Cytometry.

Specifications

Antigen Nitrotyrosine
Applications Flow Cytometry
Classification Monoclonal
Clone 39B6
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Nitrotyrosine
Host Species Mouse
Immunogen Hybridoma line 39B6
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific.
Target Species Rat, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.