Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotyrosine Antibody (EM-30), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP196130AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP196130AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP196130AI Supplier Novus Biologicals Supplier No. NBP196130AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Human, Mouse, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Nitrotyrosine
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone EM-30
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Nitrotyrosine
Host Species Mouse
Immunogen NO2-Tyr-CH2-Thyroglobulin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues
Target Species Human, Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.