Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Njmu-R1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157744
Description
Njmu-R1 Polyclonal specifically detects Njmu-R1 in Human samples. It is validated for Western Blot.Specifications
Njmu-R1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
chromosome 17 open reading frame 75, NJMU-R1, protein Njmu-R1, spermatogenesis-related protein | |
Rabbit | |
Affinity purified | |
RUO | |
64149 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9HAS0 | |
C17ORF75 | |
Synthetic peptides corresponding to Njmu-R1 The peptide sequence was selected from the N terminal of C17orf75)(50ug). Peptide sequence SSHYCLYSYRGSRLAQQRGDSEDGSPSGTNAETPSGDDFSLSLADTNLPS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Guinea pig: 92%; Equine: 92%; Canine: 85%; Rabbit: 85%; Bovine: 78%; Pig: 78%. | |
Human, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction