Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Njmu-R1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Njmu-R1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Njmu-R1 Polyclonal specifically detects Njmu-R1 in Human samples. It is validated for Western Blot.Specifications
Njmu-R1 | |
Polyclonal | |
Rabbit | |
Q9HAS0 | |
64149 | |
Synthetic peptides corresponding to Njmu-R1 The peptide sequence was selected from the N terminal of C17orf75)(50ug). Peptide sequence SSHYCLYSYRGSRLAQQRGDSEDGSPSGTNAETPSGDDFSLSLADTNLPS. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
chromosome 17 open reading frame 75, NJMU-R1, protein Njmu-R1, spermatogenesis-related protein | |
C17ORF75 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title