Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK1R Antibody, Novus Biologicals™
SDP

Catalog No. NB300101SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300101SS 0.025 mL
NB300101 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300101SS Supplier Novus Biologicals Supplier No. NB300101SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 26 publications

NK1R Polyclonal specifically detects NK1R in Guinea Pig, Human, Mouse, Rat samples. It is validated for IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen NK1R
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23792204), Immunohistochemistry-Frozen 1:1000
Formulation Whole antisera with No Preservative
Gene Accession No. P14600
Gene Alias NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor
Gene Symbols TACR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of rat Neurokinin 1 Receptor, conjugated to bovine thyroglobulin. [UniProt# P14600]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6869
Test Specificity NB300-101 is specific to the NK-1 receptor. Specificity was verified by the localization of immunohistochemical staining with NB300-101 in the neurons of mouse brain tissue.
Target Species Guinea Pig, Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.