Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK1R Rabbit anti-Human, Mouse, Rat, Clone: 9T1W2, Novus Biologicals™
SDP

Catalog No. NB168014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168014 20 μg
NB168013 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168014 Supplier Novus Biologicals Supplier No. NBP31646120UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

NK1R Monoclonal antibody specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen NK1R
Applications Western Blot
Classification Monoclonal
Clone 9T1W2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-407 of human NK1R (NK1R) (P25103).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6869
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.