Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NK1R Rabbit anti-Human, Mouse, Rat, Clone: 9T1W2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | NK1R |
---|---|
Clone | 9T1W2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
NK1R Monoclonal antibody specifically detects NK1R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
NK1R | |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6869 | |
IgG | |
Affinity purified |
9T1W2 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
NK-1R, NK1Rneurokinin 1 receptor, NKIR, SPRsubstance-P receptor, TAC1Rtachykinin 1 receptor (substance P receptor, neurokinin 1 receptor), tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor), tachykinin receptor 1NK-1 receptor | |
A synthetic peptide corresponding to a sequence within amino acids 300-407 of human NK1R (NK1R) (P25103). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title