Learn More
Description
Specifications
Specifications
| Antigen | NK3R/TACR3/Neurokinin B Receptor |
| Applications | Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Western Blot (Negative) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (PMID 30565563), Western Blot (Negative), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25496429) |
| Formulation | Whole antisera with 0.1% Sodium Azide |
| Gene Accession No. | P16177 |
| Gene Alias | Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060 |
| Gene Symbols | TACR3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
