Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK3R/TACR3/Neurokinin B Receptor Antibody, Novus Biologicals™
SDP

Catalog No. NB300102SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300102SS 0.025 mL
NB300102 0.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300102SS Supplier Novus Biologicals Supplier No. NB300102SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 31 publication

NK3R/TACR3/Neurokinin B Receptor Polyclonal specifically detects NK3R/TACR3/Neurokinin B Receptor in Guinea Pig, Human, Human (Negative), Mouse, Ovine, Rat, Sheep samples. It is validated for IF/IHC, Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Western Blot (Negative).

Specifications

Antigen NK3R/TACR3/Neurokinin B Receptor
Applications Immunocytochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Free Floating), Immunohistochemistry (Frozen), Western Blot (Negative)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500. Use reported in scientific literature (PMID 30565563), Western Blot (Negative), Immunohistochemistry Free-Floating reported in scientific literature (PMID 25496429)
Formulation Whole antisera with 0.1% Sodium Azide
Gene Accession No. P16177
Gene Alias Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060
Gene Symbols TACR3
Host Species Rabbit
Immunogen A synthetic peptide at the C-terminus of rat Neurokinin B Receptor, conjugated to bovine thyroglobulin. [UniProt# P16177]
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6870
Test Specificity NB300-102 is specific to the NK-3 receptor. Specificity was verified by the localization of immunohistochemical staining as well as pre-absorption studies with NK-3 peptide. (1)
Target Species Guinea Pig, Human, Human (Negative), Mouse, Ovine, Rat, Sheep
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.