Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK3R/TACR3/Neurokinin B Receptor Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB146599 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB146599 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB146599 Supplier Novus Biologicals Supplier No. NBP306055AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NK3R/TACR3/Neurokinin B Receptor Polyclonal specifically detects NK3R/TACR3/Neurokinin B Receptor in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen NK3R/TACR3/Neurokinin B Receptor
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NK3R/TACR3/Neurokinin B Receptor fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6870
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.