Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NK3R/TACR3/Neurokinin B Receptor Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB184743 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184743 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184743 Supplier Novus Biologicals Supplier No. NBP282076G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NK3R/TACR3/Neurokinin B Receptor Polyclonal antibody specifically detects NK3R/TACR3/Neurokinin B Receptor in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NK3R/TACR3/Neurokinin B Receptor
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Neurokinin B receptor, neurokinin beta receptor, neuromedin-K receptor, NK-3 receptor, NK-3R, NK3RMGC148061, NKR, TAC3R, TAC3RL, tachykinin receptor 3MGC148060
Host Species Rabbit
Immunogen NK3R antibody was raised against a 18 amino acid synthetic peptide from near the center of human NK3R. The immunogen is located within amino acids 370 - 420 of NK3R. Amino Acid Squence: SYDELELKTTRFHPNRQS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6870
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.