Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKCC1/SLC12A2 Antibody (915911), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346042 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346042 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346042 Supplier Novus Biologicals Supplier No. FAB8528MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKCC1/SLC12A2 Monoclonal specifically detects NKCC1/SLC12A2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen NKCC1/SLC12A2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 915911
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2, Met1-Ser1212, Accession # P55011
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6558
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.