Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKCC1/SLC12A2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10075623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10075623 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10075623 Supplier Novus Biologicals Supplier No. NB10075623

Chicken Polyclonal Antibody

NKCC1/SLC12A2 Polyclonal specifically detects NKCC1/SLC12A2 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NKCC1/SLC12A2
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P55011
Gene Alias Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2
Gene Symbols SLC12A2
Host Species Chicken
Immunogen The immunogen for this antibody was a sequence from 800a.a. to 1000a.a. from NKCC1(a 1212a.a. long protein)
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6558
Test Specificity Chicken Polyclonal to NKCC1
Target Species Human, Mouse, Rat, Bovine, Canine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.