Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2A/CD159a/KLRC1 Antibody (131411) [DyLight 405], Novus Biologicals™
SDP

Catalog No. NB401252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB401252 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB401252 Supplier Novus Biologicals Supplier No. FAB1059E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKG2A/CD159a/KLRC1 Monoclonal antibody specifically detects NKG2A/CD159a/KLRC1 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen NKG2A/CD159a/KLRC1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 131411
Conjugate DyLight 405
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD159 antigen-like family member A, CD159A, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2A and CD94
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3821
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.