Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKG2C/CD159c/KLRC2 Antibody (134522) [DyLight 680], Novus Biologicals™
SDP

Catalog No. NB400175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400175 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB400175 Supplier Novus Biologicals Supplier No. FAB1381Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NKG2C/CD159c/KLRC2 Monoclonal antibody specifically detects NKG2C/CD159c/KLRC2 in Human samples. It is validated for Flow Cytometry, CyTOF-reported

Specifications

Antigen NKG2C/CD159c/KLRC2
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 134522
Conjugate DyLight 680
Dilution Flow Cytometry, CyTOF-reported
Formulation 50mM Sodium Borate
Gene Alias CD159 antigen-like family member C, CD159c, CD159c antigen, killer cell lectin-like receptor subfamily C, member 2, MGC138244, NK cell receptor C, NKG2-C, NKG2-C type II integral membrane protein, NKG2CNKG2-C-activating NK receptor
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human NKG2C/CD159c and CD94
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3822
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.